Media Bottles
250 ml, Media Bottle
Catalog # 470205-566
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 219757
Jump to:
Product Details & Documents
Borosilicate Glass.
- Large opening, no-drip-lip, and sloping shoulders for easy pouring
- Approximate volume graduations
- Large writing area
Both the bottles and rubber-lined caps are autoclavable.
Documents
Especificaciones
- Capacity:250 ml
- Description:Media Bottle