Orders LinkContactUsLinkComponent
Media Bottles
Media Bottles
250 ml, Media Bottle
Catalog # 470205-566
Número de proveedor #:  219757
Media Bottles
Catalog # 470205-566
Media Bottles
Media Bottles
250 ml, Media Bottle
Price
$240.78
Log in Para disponibilidad
Each (1UN) Case de 48 (1UN)
Product Details & Documents
Borosilicate Glass.
  • Large opening, no-drip-lip, and sloping shoulders for easy pouring
  • Approximate volume graduations
  • Large writing area
Both the bottles and rubber-lined caps are autoclavable.
Especificaciones
  • Capacity:
    250 ml
  • Description:
    Media Bottle