Media Bottles
125 ml, Media Bottle
470205-564
: 219755
Borosilicate Glass.
- Large opening, no-drip-lip, and sloping shoulders for easy pouring
- Approximate volume graduations
- Large writing area
Both the bottles and rubber-lined caps are autoclavable.
- Capacity:125 ml
- Description:Media Bottle