XXXX Quality Corks
7
470204-538
: 50724
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
- Fine Grain
- Ideal for Use as Tight-Fitting Stoppers
- Common Sizes
- Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
: Product contains latex. May cause allergic reactions.
- Stopper No.:7
- Cat. No.:470204-538
- Environmentally Preferable:Low Manufacturing Impact