Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Quantitative Grade Filter Paper Circles

VWR® Quantitative Grade Filter Paper Circles

Quantitative Grade Filter Paper Circles, VWR® Grade #494, Diameter: 9 cm
Catalog # 470204-444
VWR® Quantitative Grade Filter Paper Circles
Catalog # 470204-444
Supplier:  
VWR® Quantitative Grade Filter Paper Circles
VWR® Quantitative Grade Filter Paper Circles

Quantitative Grade Filter Paper Circles, VWR® Grade #494, Diameter: 9 cm

Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
FILTER PAPER #2 9CM PK100 Catalog #28455-085

El producto 28455-085 es un reemplazo para 470204-444 y tiene el mismo embalaje y especificaciones, pero puede tener una cantidad diferente.

Product Details & Documents

Quantitative filter papers are used for gravimetric analyses or filtrations where the precipitate is to be recovered.

  • Range of porosity available for filtering out precipitates
  • Flow rate varies between models

Sargent-Welch Quantitative

VWR® Grade #494
Porosity: Fine - Ashless
Flow Rate: Slow
Particle Retention: 1 μm
Applications: Useful for very fine precipitates. Ideal for BaSO₄, PbSO₄, SnS2, NiS, CaOH₂

VWR® Grade #474
Porosity: Medium - Ashless
Flow Rate: Medium
Particle Retention: 5 to 10 μm
Applications: For general quantitative analysis. Ideal for the filtration of AlPO₄, ZnNH₄PO₄, Ag₂CrO₄

Especificaciones
  • Description:
    Quantitative Grade Filter Paper Circles, VWR® Grade #494, Diameter: 9 cm