XXXX Quality Corks
6
Catalog # 470204-426
Supplier: MANTON INDUSTRIAL CORK SE
Supplier #: 50624
XXXX Quality Corks
Catalog # 470204-426
Supplier: MANTON INDUSTRIAL CORK SE
Jump to:
Product Details & Documents
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
- Fine Grain
- Ideal for Use as Tight-Fitting Stoppers
- Common Sizes
- Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
Caution: Product contains latex. May cause allergic reactions.
Specifications
- Stopper No.:6
- Cat. No.:470204-426
- Environmentally Preferable:Low Manufacturing Impact