Orders LinkContactUsLinkComponent
XXXX Quality Corks
XXXX Quality Corks
6
Catalog # 470204-426
   Low Manufacturing Impact
Supplier #:  50624
XXXX Quality Corks
Catalog # 470204-426
   Low Manufacturing Impact
XXXX Quality Corks
XXXX Quality Corks
6
Price
$222.80
Log in for availability
Paquete de 100
Product Details & Documents
XXXX Quality Corks have a fine grain and are ideal for use as tight-fitting stoppers.
  • Fine Grain
  • Ideal for Use as Tight-Fitting Stoppers
  • Common Sizes
  • Regular Length
The listed sizes listed are common in school science laboratories.
The corks are regular length.
Caution: Product contains latex. May cause allergic reactions.
Specifications
  • Stopper No.:
    6
  • Cat. No.:
    470204-426
  • Environmentally Preferable:
    Low Manufacturing Impact