Scheibler Desiccators
Scheibler Desiccators, ID, mm: 250
Catalog # 470204-328
Jump to:
Discontinued
Product Details & Documents
Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly. Porcelain plates sold separately.
Specifications
- Description:Scheibler Desiccators, ID, mm: 250