Scheibler Desiccator
Scheibler Desiccator, 200 mm, —, —, —
470204-326
: 682-4
Non-Vacuum Desiccator
- Made of heavy, flint glass
Made of clear glass with thick uniform walls and a heavy, broad flange. Cover with molded knob is ground to fit the flange perfectly.
- Hole Ø:—
- Ø int.:200 mm
- No. of Holes:—
- Description:Scheibler Desiccator
- Plate Diameter:—