Order Entry
Mexico
Orders LinkContactUsLinkComponent
Test Tube Brushes

Test Tube Brushes

For tube diameter up to 16 mm, Natural bristle, 89 mm, 19 mm (3/4"), 229 mm
Catalog # 470203-400
Supplier:   MILL-ROSE COMPANY
Supplier #:  72002W
Test Tube Brushes
Catalog # 470203-400
Supplier:  MILL-ROSE COMPANY
Test Tube Brushes
Test Tube Brushes

For tube diameter up to 16 mm, Natural bristle, 89 mm, 19 mm (3/4"), 229 mm

Price
$31.17
Log in for availability
Each (1UN) Paquete de 12 (1UN)
Product Details & Documents

Will Not Scratch.

  • Choose between natural or nylon bristles
  • Neither the bristles or the radial end will scratch glass
Specifications
  • Bristle Material:
    Natural bristle
  • Brush head Ø:
    19 mm (3/4")
  • Description:
    For tube diameter up to 16 mm
  • Total length:
    229 mm
  • Brush Head Length:
    89 mm