Orders LinkContactUsLinkComponent
Hydrion pH Test Papers
pH paper roll, 6.0 - 8.0
Catalog # 470203-380
 :  445
Hydrion pH Test Papers
Catalog # 470203-380
Hydrion pH Test Papers
Hydrion pH Test Papers
pH paper roll, 6.0 - 8.0
Price
$800.26
Log in Para disponibilidad
Paquete de 5 Cartón de 10 (5UN)
Product Details & Documents
Includes Color Comparison Chart.
  • Wide range shows sharp color change for each full ph unit
  • Narrow range papers show a color change for each half pH unit
The wide range (1.0 - 12.0 and 1.0 - 14.0) shows a sharp color change for each full pH unit. The narrow range papers show a color change for each half pH unit. Packages of five rolls.
Especificaciones
  • Description:
    pH paper roll
  • pH Range:
    6.0 - 8.0