Hydrion pH Test Papers
pH paper roll, 6.0 - 8.0
Catalog # 470203-380
Supplier: MICRO ESSENTIAL LABORATORY I
: 445
Jump to:
Product Details & Documents
Includes Color Comparison Chart.
- Wide range shows sharp color change for each full ph unit
- Narrow range papers show a color change for each half pH unit
The wide range (1.0 - 12.0 and 1.0 - 14.0) shows a sharp color change for each full pH unit. The narrow range papers show a color change for each half pH unit. Packages of five rolls.
Especificaciones
- Description:pH paper roll
- pH Range:6.0 - 8.0