Conductivity/TDS Calibration Solution
Conductivity/TDS Calibration Solution, 718 μS, 503 ppm
Supplier: LA MOTTE CHEMICAL PROD CO SE
Número de proveedor #: 6417-L
Conductivity/TDS Calibration Solution
Catalog # 470203-120
Supplier: LA MOTTE CHEMICAL PROD CO SE
CAS Number:
Jump to:
Product Details & Documents
Always keep your meter in range.
- Already prepared solutions
- Various ranges
- Aligned with GLOBE protocols
Prepared solution to keep your conductivity calibrated. 500ml bottles.
Documents
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Description:Conductivity/TDS Calibration Solution
- Conductivity:718 μS
- Dissolved Solids:503 ppm
- Cat. No.:470203-120