Test Tube Brushes
For tube diameter 9.5-12.5 mm, Natural bristle
470202-904
: J610124
These test tube brushes feature a radially tufted tip.
- Heavy, Galvanized Wire
- Radially Tufted Tip
- Nylon or Natural Brushes Available
They also feature heavy, galvanized wire.
Nylon or natural bristles are available.
- Bristle Material:Natural bristle
- Description:For tube diameter 9.5-12.5 mm