Order Entry
Mexico
Orders LinkContactUsLinkComponent
Electric Heating Mantles

Electric Heating Mantles

100 ml, 80 W
Catalog # 470202-744
Número de proveedor #:  100A O396
Electric Heating Mantles
Catalog # 470202-744
Electric Heating Mantles
Electric Heating Mantles

100 ml, 80 W

Price
$8,037.77
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Designed to Operate Below Red Heat at Maximum Input Voltage.

  • Made entirely of glass fabric
  • Hemispherical construction allows full view of flask contents while heating

Mantles provide evenly distributed heat which may be closely controlled by adjustment of the input voltage. (Heating Mantle Controller sold separately WLS-40883 / 470202-750)
With separable 2-wire line cord, 4' L. For 115VAC or DC. CSA listed.

Certifications: CSA listed.

Especificaciones
  • Power:
    80 W
  • Flask Volume:
    100 ml