Orders LinkContactUsLinkComponent
Wooden Slide Boxes
Wooden Slide Boxes
Slide Box, Wooden, 100, 23 x 19 x 3 cm
Catalog # 470201-936
Supplier #:  3304
Wooden Slide Boxes
Catalog # 470201-936
Wooden Slide Boxes
Wooden Slide Boxes
Slide Box, Wooden, 100, 23 x 19 x 3 cm
Price
$1,452.45
Log in for availability
Each (1UN)
Product Details & Documents
Excellent For Storing Standard Sized Slides
  • Keep slides debris-free
  • Store slides neatly and efficiently
These high quality, wooden slide boxes hold standard slides in individual slots. Supplied with an index for recording slide titles.
Specifications
  • Capacity:
    100
  • Description:
    Slide Box, Wooden
  • Dimensions:
    23 x 19 x 3 cm