Flask Brushes
White, Plastic, 14"
Catalog # 470201-502
Supplier: ALLIED K & R
Número de proveedor #: D131
Jump to:
Product Details & Documents
White bristle and plastic handle.
- Multiple sizes available
Recommended for volumetric and boiling flasks because of the unusally flexible, curved steel spring that enables the brush to reach all parts of the flask.
Especificaciones
- Bristle Color:White
- Handle Material:Plastic
- Total length:14"