Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Quantitative Grade Filter Paper Circles

VWR® Quantitative Grade Filter Paper Circles

Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 7 cm
Catalog # 470201-458
Número de proveedor #:  0540-0700
VWR® Quantitative Grade Filter Paper Circles
Catalog # 470201-458
VWR® Quantitative Grade Filter Paper Circles
VWR® Quantitative Grade Filter Paper Circles

Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 7 cm

Price
$628.93
Log in Para disponibilidad
Paquete de 100
Product Details & Documents

Quantitative filter papers are used for gravimetric analyses or filtrations where the precipitate is to be recovered.

  • Range of porosity available for filtering out precipitates
  • Flow rate varies between models

Sargent-Welch Quantitative

VWR® Grade #494
Porosity: Fine - Ashless
Flow Rate: Slow
Particle Retention: 1 μm
Applications: Useful for very fine precipitates. Ideal for BaSO₄, PbSO₄, SnS2, NiS, CaOH₂

VWR® Grade #474
Porosity: Medium - Ashless
Flow Rate: Medium
Particle Retention: 5 to 10 μm
Applications: For general quantitative analysis. Ideal for the filtration of AlPO₄, ZnNH₄PO₄, Ag₂CrO₄

Especificaciones
  • Description:
    Quantitative Grade Filter Paper Circles, Sargent-Welch Quantitative, Diameter: 7 cm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.