Polyethylene Dispensing Bottles
470201-238
: F11637-0008_SE
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.
Low-density polyethylene bottles and spouts with sealer caps that snap on securely. Spout openings are standard 0.030" and can be enlarged with scissors.