VWR® Linear Spring Scales
250 g x 5 g (2.5 N x 0.05 N)
Catalog # 470201-018
Supplier: SHIVDIAL SUD & SONS
Número de proveedor #: PH10120/2
Jump to:
Product Details & Documents
Designed for Heavy Duty Student Use
- Feature easy to read markings
- Durable for years of regular student use
- Length: 14 cm
- Each scale is easily zeroed
Each has a plated steel suspension ring at the top, and a hook at the bottom.
Especificaciones
- Weighing Capacity:250 g x 5 g (2.5 N x 0.05 N)