Laboratory Magnet Set
Laboratory Magnet Set
Catalog # 470200-924
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MSET4
Jump to:
Product Details & Documents
A must for the Lab
- Set of four
The magnets in this set are permanent Alnico and include two bars, two horseshoes, and keepers. Sizes: bars 3"L x 5/8"W x 3/8"H; small horseshoe 1 "W x 1"H x 3/8"D; large horseshoe 1 3/8"W x 1 3/8"H x 3/4"D.
Especificaciones
- Description:Laboratory Magnet Set