Squibb Urinometer and Jar
Midget Urinometer
Catalog # 470200-844
Supplier: VWR International
Jump to:
Product Details & Documents
Perfect for when only minimal volumes are required, this urinometer provides highly accurate samples.
- Calibrated, Easy-to-Read Scale
- Range: 1.000–1.060 Sp. Gr
- Accurate Samples
- Stain-Free Clear Glass
- Capacity: 50 mL
This stain-free and clear glass apparatus has a capacity of 50 mL and a wide base that prevents it from tipping.
Its range is 1.000–1.060 Sp. Gr in 0.002 graduations.
Specifications
- Description:Midget Urinometer