Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
El producto 470002-478 es un reemplazo para 470200-078 y tiene el mismo embalaje y especificaciones, pero puede tener una cantidad diferente.
Product Details & Documents
This kit explores chemical nomenclature through experimentation.
- Ideal for Grades 7-10
- Students Learn to Recognize Evidence of Chemical Changes
- Kit Illustrates Concepts of Elements, Compounds, and Mixtures
- Special Emphasis on Creating New Compounds in Chemical Reactions
- Enough Material to Repeat Experiment Twice
This kit illustrates the concepts of elements, compounds, and mixtures with special emphasis on creating new compounds in chemical reactions. Students witness evidence of chemical changes identified by the formation of precipitates, color changes, odors, or the production of gas.
The kit includes enough material to repeat the experiment twice. Worksheets focus on chemical symbols, chemical formulas, chemical names, components of common compounds, and distinguishing between chemical and physical changes.
Ordering information: This lab includes an instructor manual, a SDS, filter paper, spoons, calcium chloride, sodium carbonate, copper sulfate, and sodium bicarbonate. Balance and laboratory supplies are needed but not included.
Especificaciones
- Description:Elements, Compounds, and Mixtures II Lab Activity
- No. of Lab Groups:6
- Includes:Instructions and Student Worksheets
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.