Orders LinkContactUsLinkComponent
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Elements, Compounds, and Mixtures II Lab Activity, Instructions and Student Worksheets, 6
Catalog # 470200-078
Número de proveedor #:  13-108
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Catalog # 470200-078
Supplier:  
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Kemtec™ Elements, Compounds, and Mixtures II Lab Activity
Elements, Compounds, and Mixtures II Lab Activity, Instructions and Student Worksheets, 6
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
LAB KIT ELEMENTS COMPOUNDS MIXTURES II Catalog #470002-478
El producto 470002-478 es un reemplazo para 470200-078 y tiene el mismo embalaje y especificaciones, pero puede tener una cantidad diferente.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit explores chemical nomenclature through experimentation.
  • Ideal for Grades 7-10
  • Students Learn to Recognize Evidence of Chemical Changes
  • Kit Illustrates Concepts of Elements, Compounds, and Mixtures
  • Special Emphasis on Creating New Compounds in Chemical Reactions
  • Enough Material to Repeat Experiment Twice
This kit illustrates the concepts of elements, compounds, and mixtures with special emphasis on creating new compounds in chemical reactions. Students witness evidence of chemical changes identified by the formation of precipitates, color changes, odors, or the production of gas.
The kit includes enough material to repeat the experiment twice. Worksheets focus on chemical symbols, chemical formulas, chemical names, components of common compounds, and distinguishing between chemical and physical changes.
Ordering information: This lab includes an instructor manual, a SDS, filter paper, spoons, calcium chloride, sodium carbonate, copper sulfate, and sodium bicarbonate. Balance and laboratory supplies are needed but not included.
Especificaciones
  • Description:
    Elements, Compounds, and Mixtures II Lab Activity
  • No. of Lab Groups:
    6
  • Includes:
    Instructions and Student Worksheets