Jar with Cap, HDPE
Jar with Cap, HDPE, 120 mL
Catalog # 470191-496
Supplier: DYNALON
Número de proveedor #: 226415-0120
Jump to:
Product Details & Documents
These cylindrical food-grade HDPE containers with black HDPE screw closures are supplied with a neck insert for added security, suitable for powders, solid specimens, creamy or semisolid substances, crystals, and hygroscopic substances.
- Aligned with GLOBE protocols
Especificaciones
- Description:Jar with Cap, HDPE, 120 mL