Orders LinkContactUsLinkComponent
Jar with Cap, HDPE
Jar with Cap, HDPE
Jar with Cap, HDPE, 120 mL
Catalog # 470191-496
Supplier:   DYNALON
Número de proveedor #:  226415-0120
Jar with Cap, HDPE
Catalog # 470191-496
Supplier:  DYNALON
Jar with Cap, HDPE
Jar with Cap, HDPE
Jar with Cap, HDPE, 120 mL
Price
$101.59
Log in Para disponibilidad
Each (1UN) Paquete de 10 (1UN)
Product Details & Documents
These cylindrical food-grade HDPE containers with black HDPE screw closures are supplied with a neck insert for added security, suitable for powders, solid specimens, creamy or semisolid substances, crystals, and hygroscopic substances.
  • Aligned with GLOBE protocols
Especificaciones
  • Description:
    Jar with Cap, HDPE, 120 mL