Orders LinkContactUsLinkComponent
Polyethylene Wash Bottles
125 ml
  470191-312
Polyethylene Wash Bottles
  470191-312
Polyethylene Wash Bottles
Polyethylene Wash Bottles
125 ml
$25.79
Each (1UN) Case de 48 (1UN)
Unbreakable wash bottles for student use.
  • Polyethylene, narrow mouth construction
  • Closures included
Cap and delivery tube are one-piece construction to eliminate leakage. The inside diameter of the delivery tip is carefully drawn to give a fine stream of liquid.
  • Capacity:
    125 ml