125 ml
Unbreakable wash bottles for student use.
Cap and delivery tube are one-piece construction to eliminate leakage. The inside diameter of the delivery tip is carefully drawn to give a fine stream of liquid.
#: 470005-894
: VWR International