Orders LinkContactUsLinkComponent
Polyethylene Wash Bottles
250 ml
  470191-302
Polyethylene Wash Bottles
  470191-302
Polyethylene Wash Bottles
Polyethylene Wash Bottles
250 ml
$32.26
Each (1UN) Case de 36 (1UN)
Unbreakable wash bottles for student use.
  • Polyethylene, narrow mouth construction
  • Closures included
Cap and delivery tube are one-piece construction to eliminate leakage. The inside diameter of the delivery tip is carefully drawn to give a fine stream of liquid.
  • Capacity:
    250 ml