Polyethylene Wash Bottles
500 ml
Supplier: VWR International
Jump to:
Product Details & Documents
Unbreakable wash bottles for student use.
- Polyethylene, narrow mouth construction
- Closures included
Cap and delivery tube are one-piece construction to eliminate leakage. The inside diameter of the delivery tip is carefully drawn to give a fine stream of liquid.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:500 ml