VWR® Volumetric Flasks
1000 ml, Snap cap
Supplier: VWR International
Jump to:
Product Details & Documents
A Great Value!
- Class B borosilicate glass
- Includes choice of glass stopper or snap cap
- Graduation line for accurate measurements
Our new Class B borosilicate volumetric flasks feature capacity tolerances twice those specified for Class A volumetric ware.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:1000 ml
- Stopper Type:Snap cap