Orders LinkContactUsLinkComponent
VWR® Volumetric Flasks
VWR® Volumetric Flasks
100 ml, Snap cap
Catalog # 470191-256
Supplier:   VWR International
VWR® Volumetric Flasks
Catalog # 470191-256
Supplier:  VWR International
VWR® Volumetric Flasks
VWR® Volumetric Flasks
100 ml, Snap cap
Price
$103.72
Log in for availability
Each (1UN) Case de 12 (1UN)
Product Details & Documents
A Great Value!
  • Class B borosilicate glass
  • Includes choice of glass stopper or snap cap
  • Graduation line for accurate measurements
Our new Class B borosilicate volumetric flasks feature capacity tolerances twice those specified for Class A volumetric ware.
Documents
Specifications
  • Capacity:
    100 ml
  • Stopper Type:
    Snap cap