VWR® Florence Boiling Flasks
1000 ml
Supplier: VWR International
Jump to:
Product Details & Documents
Our high-quality borosilicate Florence flasks meet ASTM standards and are a great value.
- Boil liquids without excessive rapid evaporation
- High-quality borosilicate glass fabrication
Their round body and long neck are ideal for boiling liquids without excessively rapid evaporation.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:1000 ml