Orders LinkContactUsLinkComponent
VWR® Cylinders with Plastic Base
VWR® Cylinders with Plastic Base
100 ml
Catalog # 470191-194
Supplier:   VWR International
VWR® Cylinders with Plastic Base
Catalog # 470191-194
Supplier:  VWR International
VWR® Cylinders with Plastic Base
VWR® Cylinders with Plastic Base
100 ml
Price
$113.40
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
  • Double graduated metric scale
  • Pour-out
  • Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.
Documents
Especificaciones
  • Capacity:
    100 ml
  • Cat. No.:
    470191-194
  • Form:
    Tall