VWR® Cylinders with Plastic Base
100 ml
Supplier: VWR International
Jump to:
Product Details & Documents
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
- Double graduated metric scale
- Pour-out
- Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:100 ml
- Cat. No.:470191-194
- Form:Tall