Orders LinkContactUsLinkComponent
VWR® Cylinders with Plastic Base
50 ml
  470191-192
VWR® Cylinders with Plastic Base
  470191-192
VWR® Cylinders with Plastic Base
VWR® Cylinders with Plastic Base
50 ml
$68.80
Each (1UN) Case de 12 (1UN)
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
  • Double graduated metric scale
  • Pour-out
  • Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.
  • Capacity:
    50 ml
  • Cat. No.:
    470191-192
  • Form:
    Tall