Orders LinkContactUsLinkComponent
VWR® Cylinders with Plastic Base
VWR® Cylinders with Plastic Base
10 ml
Catalog # 470191-190
Supplier:   VWR International
VWR® Cylinders with Plastic Base
Catalog # 470191-190
Supplier:  VWR International
VWR® Cylinders with Plastic Base
VWR® Cylinders with Plastic Base
10 ml
Price
$48.90
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Our high-quality borosilicate cylinders meet ASTM standards and are a great value.
  • Double graduated metric scale
  • Pour-out
  • Detachable plastic base
Featuring uniform white enamel, they can be heated and used with acids, salt solutions, and organic solvents.
Documents
Especificaciones
  • Capacity:
    10 ml
  • Cat. No.:
    470191-190
  • Form:
    Tall