Orders LinkContactUsLinkComponent
VWR® Volumetric Flasks
500 ml, Glass Stopper
  470190-862
VWR® Volumetric Flasks
  470190-862
VWR® Volumetric Flasks
VWR® Volumetric Flasks
500 ml, Glass Stopper
$263.89
Each (1UN) Case de 12 (1UN)
A Great Value!
  • Class B borosilicate glass
  • Includes choice of glass stopper or snap cap
  • Graduation line for accurate measurements
Our new Class B borosilicate volumetric flasks feature capacity tolerances twice those specified for Class A volumetric ware.
  • Capacity:
    500 ml
  • Stopper Type:
    Glass Stopper