Coplin Staining Jars
Up to 10 Slides, Coplin Staining Jar, Plastic
Catalog # 470174-652
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: 62101-PK/12
Jump to:
Product Details & Documents
Coplin Jar with Screw-on Top Prevents Spills.
- Quality slide making supplies
- Slide staining tools
The screw-on cover prevents evaporation and contamination when staining. Each jar holds five single slides or ten back to back.
Especificaciones
- Capacity:Up to 10 Slides
- Description:Coplin Staining Jar, Plastic