Cells: Structure, Function and Processes Posters
Cells Poster Set, Laminated, 61 cm, 46 cm
Catalog # 470161-168
Supplier: JAGUAR EDUCATIONAL SE
Número de proveedor #: 03-PS05L-B
Cells: Structure, Function and Processes Posters
Catalog # 470161-168
Supplier: JAGUAR EDUCATIONAL SE
Jump to:
Product Details & Documents
Zoom in on the key topics in cell biology with these colorful and innovative posters.
- Beautifully illustrated
- Highly detailed
Labeled diagrams and descriptive text highlight important structures and processes. Guaranteed to draw attention, the ten laminated posters include: Animal Cell, Meiosis, Plant Cell, Bacteria Cell, Mitosis and Cytokinesis, Cellular Respiration, Active and Passive Transport, DNA and RNA, Photosynthesis and Cell Specialization. Set of 10.
Especificaciones
- Width:46 cm
- Description:Cells Poster Set
- Length:61 cm
- Finish:Laminated
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



