Orders LinkContactUsLinkComponent
Fast Release Pi-Pumps
Fast Release Pi-Pumps
Fast Release Pi-Pump, Green, 10 ml
Catalog # 470153-804
Supplier:   BEL-ART PRODUCTS SE
Número de proveedor #:  F37904-0010
Fast Release Pi-Pumps
Catalog # 470153-804
Fast Release Pi-Pumps
Fast Release Pi-Pumps
Fast Release Pi-Pump, Green, 10 ml
Price
$976.00
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents
Designed for fast and accurate one-handed operation.
  • Reduce the risk of injury from repetitive pipetting
  • Expel entire contents in one shot or smaller amounts precisely
Each pump is resistant to acids, bases, and most solvents; and it can be easily disassembled for cleaning.
Especificaciones
  • Capacity:
    10 ml
  • Description:
    Fast Release Pi-Pump, Green