Fast Release Pi-Pumps
Fast Release Pi-Pump, Green, 10 ml
Catalog # 470153-804
Supplier: BEL-ART PRODUCTS SE
Número de proveedor #: F37904-0010
Jump to:
Product Details & Documents
Designed for fast and accurate one-handed operation.
- Reduce the risk of injury from repetitive pipetting
- Expel entire contents in one shot or smaller amounts precisely
Each pump is resistant to acids, bases, and most solvents; and it can be easily disassembled for cleaning.
Especificaciones
- Capacity:10 ml
- Description:Fast Release Pi-Pump, Green