Fast Release Pi-Pumps
Fast Release Pi-Pump, 10 ml
Catalog # 470145-290
: F37904-0010
Jump to:
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
Product Details & Documents
Designed for fast and accurate one-handed operation, this pump will reduce the risk of injury from repetitive pipetting.
- Each pump is resistant to acids, bases, and most solvents
- Disassembly for cleaning is easy
The fast-release trigger expels the entire contents at one shot, or you can use the thumb wheel for precision dispensing.
Especificaciones
- Capacity:10 ml
- Description:Fast Release Pi-Pump