Orders LinkContactUsLinkComponent
Canonical URL fetch failed:ERROR
Balls for Physics Experiments
Drilled Brass Ball, 25 mm, 1"
Catalog # 470138-848
Balls for Physics Experiments
Catalog # 470138-848
Supplier:  
Balls for Physics Experiments
Balls for Physics Experiments
Drilled Brass Ball, 25 mm, 1"
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BALL BRASS DRILLED 1IN / 25MM D Catalog #470007-150
Product 470007-150 is a replacement for 470138-848 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Ideal for Collision and Pendulum Demonstrations
Drilled balls for pendulum bobs or impact experiments. 
Specifications
  • Description:
    Drilled Brass Ball, 25 mm
  • Diameter:
    1"