Glue Gun
Glue Sticks, 7/16"
Catalog # 470101-400
Supplier: NOTIONS MARKETING
Número de proveedor #: DT-6
Jump to:
Product Details & Documents
This is a low temperature mini glue gun.
- Easy to handle
- No drip nozzle
- Built-in stand
A low-temperature glue gun is much safer to use because it decreases the likelihood of burns. Great for wood, plastic, fabric, metal, and paper. Glue sticks are available separately. 120 VAC.
Especificaciones
- Description:Glue Sticks, 7/16"