Ward's® Diopside
Diopside, Hand Specimen, 2×3" - 3×4"
Catalog # 470025-744
Supplier: Avantor
Jump to:
Product Details & Documents
Pyroxene group; gray-green, coarse crystalline.
- WARD’S offers different specimen sizes to meet your various needs
- Student specimens, hand specimens, and bulk packs are available
Especificaciones
- Description:Diopside, Hand Specimen
- Specimen Dimensions:2×3" - 3×4"