Orders LinkContactUsLinkComponent
Ward's® Diopside
Diopside, Bulk Pack, 1 kg, Size will vary
  470025-742
 :   Avantor
Ward's® Diopside
  470025-742
 :  Avantor
Ward's® Diopside
Ward's® Diopside
Diopside, Bulk Pack, 1 kg, Size will vary
$506.88
Each (1UN)
Pyroxene group; gray-green, coarse crystalline.
  • WARD’S offers different specimen sizes to meet your various needs
  • Student specimens, hand specimens, and bulk packs are available
  • Description:
    Diopside, Bulk Pack, 1 kg
  • Specimen Dimensions:
    Size will vary