Ward's® Diopside
Diopside, Student Specimens, 1×1" - 1×2"
Catalog # 470025-740
Supplier: Avantor
Jump to:
Product Details & Documents
Pyroxene group; gray-green, coarse crystalline.
- WARD’S offers different specimen sizes to meet your various needs
- Student specimens, hand specimens, and bulk packs are available
Especificaciones
- Description:Diopside, Student Specimens
- Specimen Dimensions:1×1" - 1×2"