Orders LinkContactUsLinkComponent
Ward's® Labradorite (Cleavage)
Ward's® Labradorite (Cleavage)
Labradorite (Cleavage), Hand Specimen, 2×3" - 3×4"
Catalog # 470025-662
Supplier:   Avantor
Ward's® Labradorite (Cleavage)
Catalog # 470025-662
Supplier:  Avantor
Ward's® Labradorite (Cleavage)
Ward's® Labradorite (Cleavage)
Labradorite (Cleavage), Hand Specimen, 2×3" - 3×4"
Price
$376.30
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Feldspar group; good cleavages of pure soda lime feldspar; most show twinning and labradorescence.
  • Collected in Quebec
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Labradorite (Cleavage), Hand Specimen
  • Specimen Dimensions:
    2×3" - 3×4"