Orders LinkContactUsLinkComponent
Ward's® Kaolin (Massive)
Ward's® Kaolin (Massive)
Kaolin (Massive), Student Specimens, 1×1 - 1×1"
Catalog # 470025-652
Supplier:   Avantor
Ward's® Kaolin (Massive)
Catalog # 470025-652
Supplier:  Avantor
Ward's® Kaolin (Massive)
Ward's® Kaolin (Massive)
Kaolin (Massive), Student Specimens, 1×1 - 1×1"
Price
$294.40
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Pure, white, earthy “China clay”.
  • Collected in Georgia
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Kaolin (Massive), Student Specimens
  • Specimen Dimensions:
    1×1 - 1×1"
  • Cat. No.:
    470025-652