Orders LinkContactUsLinkComponent
Ward's® Hematite (Specular)
Hematite (Specular), Student Specimens, 1×1" - 1×1"
  470025-642
 :   Avantor
Ward's® Hematite (Specular)
  470025-642
 :  Avantor
Ward's® Hematite (Specular)
Ward's® Hematite (Specular)
Hematite (Specular), Student Specimens, 1×1" - 1×1"
$301.25
Paquete de 10
Black, Micaceous, glittery metallic.
  • Collected in Michigan
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
  • Description:
    Hematite (Specular), Student Specimens
  • Specimen Dimensions:
    1×1" - 1×1"