Orders LinkContactUsLinkComponent
Galena
Galena
Galena, hand specimen, —
Catalog # 470025-588
Supplier:   Avantor
Galena
Catalog # 470025-588
Supplier:  Avantor
Galena
Galena
Galena, hand specimen, —
Price
$536.08
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Bright metallic cubic cleavages; lead ore.
  • Collected in Morocco
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Galena, hand specimen
  • Specimen Dimensions:
Recommendations