Orders LinkContactUsLinkComponent
Galena
Galena bulk pack, 1 kg, —
  470025-586
 :   Avantor
Galena
  470025-586
 :  Avantor
Galena
Galena
Galena bulk pack, 1 kg, —
$591.63
Each (1UN)
Bright metallic cubic cleavages; lead ore.
  • Collected in Morocco
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
  • Description:
    Galena bulk pack, 1 kg
  • Specimen Dimensions:
Recommendations