Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Dolomite (Coarse)

Ward's® Dolomite (Coarse)

Dolomite (Coarse), bulk pack, 1 kg, Size will vary
Catalog # 470025-558
Supplier:   Avantor
Ward's® Dolomite (Coarse)
Catalog # 470025-558
Supplier:  Avantor
Ward's® Dolomite (Coarse)
Ward's® Dolomite (Coarse)

Dolomite (Coarse), bulk pack, 1 kg, Size will vary

Price
$290.38
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

Coarse, 'salt and pepper': dark pyroxene and light andesine. Provides perfect representation of an intermediate igneous rock.

  • Collected in New Jersey

  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available

Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.

Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.

Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.

Especificaciones
  • Description:
    Dolomite (Coarse), bulk pack, 1 kg
  • Specimen Dimensions:
    Size will vary
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.