Orders LinkContactUsLinkComponent
Ward's® Dolomite (Coarse)
Ward's® Dolomite (Coarse)
Dolomite (Coarse), student specimens, 1×1 - 1×1"
Catalog # 470025-556
Supplier:   Avantor
Ward's® Dolomite (Coarse)
Catalog # 470025-556
Supplier:  Avantor
Ward's® Dolomite (Coarse)
Ward's® Dolomite (Coarse)
Dolomite (Coarse), student specimens, 1×1 - 1×1"
Price
$214.78
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Coarse, 'salt and pepper': dark pyroxene and light andesine. Provides perfect representation of an intermediate igneous rock.
  • Collected in New Jersey
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Dolomite (Coarse), student specimens
  • Specimen Dimensions:
    1×1 - 1×1"