Ward's® Calcite (Cleavage)
Calcite (Cleavage), student specimen, 1×1 - 1×1"
470025-510
: Avantor
Pure translucent to white, rhombic cleavages, Mohs’ hardness of 3.
Collected in Mexico
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
- Description:Calcite (Cleavage), student specimen
- Specimen Dimensions:1×1 - 1×1"