Orders LinkContactUsLinkComponent
Ward's® Calcite (Cleavage)
Calcite (Cleavage), student specimen, 1×1 - 1×1"
  470025-510
 :   Avantor
Ward's® Calcite (Cleavage)
  470025-510
 :  Avantor
Ward's® Calcite (Cleavage)
Ward's® Calcite (Cleavage)
Calcite (Cleavage), student specimen, 1×1 - 1×1"
$344.80
Paquete de 10
Pure translucent to white, rhombic cleavages, Mohs’ hardness of 3.
  • Collected in Mexico
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
  • Description:
    Calcite (Cleavage), student specimen
  • Specimen Dimensions:
    1×1 - 1×1"