Ward's® Testing Your Sense Organs Lab Activity
Learn The Capabilities And Limitations Of The Sense Organs
- Materials For 15 Setups
- Links To Real World Concepts
- Estimated Class/Lab Time Required: 45 Minutes
Students perform a battery of tests to determine the responsiveness of their own senses, including blind spot and after-image testing, flavor discrimination, taste bud mapping, thermoreceptor mapping, sensory adaptation testing, and more!
: A refill kit is available and contains only consumable component items. It does not contain additional student or teacher guides.
- Description:Testing Your Sense Organs Refill
- No. of Lab Groups:15