Orders LinkContactUsLinkComponent
Ward's® Testing Your Sense Organs Lab Activity
Testing Your Sense Organs Lab Activity, 15
  470023-814
 :   Avantor
Ward's® Testing Your Sense Organs Lab Activity
  470023-814
 :  Avantor
Ward's® Testing Your Sense Organs Lab Activity
Ward's® Testing Your Sense Organs Lab Activity
Testing Your Sense Organs Lab Activity, 15
$2,229.00
Kit de 1
Learn The Capabilities And Limitations Of The Sense Organs
  • Materials For 15 Setups
  • Links To Real World Concepts
  • Estimated Class/Lab Time Required: 45 Minutes
Students perform a battery of tests to determine the responsiveness of their own senses, including blind spot and after-image testing, flavor discrimination, taste bud mapping, thermoreceptor mapping, sensory adaptation testing, and more!
 : A refill kit is available and contains only consumable component items. It does not contain additional student or teacher guides.
  • Description:
    Testing Your Sense Organs Lab Activity
  • No. of Lab Groups:
    15
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.